Methyl-m-anisat 98%

Lieferant: Apollo Scientific

Synonyme: Methyl-3-methoxybenzoat, 3-Anissäuremethylester, 3-Methoxybenzoesäuremethylester, m-Anissäuremethylester

OR914552-100G
APOSOR914552-100GEA 77 EUR
APOSOR914552-100G
Methyl-m-anisat 98%
Methyl-m-anisat
Formel: C₉H₁₀O₃
Molecular Weight: 166,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00017193
CAS-Nummer: 5368-81-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR