1-Methyl-1H-indole-5-carboxylic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Methyl-5-indolecarboxylic Acid
OR9146-1G
OR9146-250MG
APOSOR9146-1GEA
98.6
EUR
APOSOR9146-1G
APOSOR9146-250MG
1-Methyl-1H-indole-5-carboxylic acid 97%
1-Methyl-1H-indole-5-carboxylic acid
Formel:
C₁₀H₉NO₂ Molecular Weight: 175,19 g/mol Schmelzpunkt: 221…223 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03839859 CAS-Nummer: 186129-25-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...