2-Nitrobenzylamine hydrochloride 96%
Lieferant: Apollo Scientific
Synonyme: (2-Nitrophenyl)methanamine hydrochloride
OR914657-5G
APOSOR914657-5GEA 66.7 EUR
APOSOR914657-5G
2-Nitrobenzylamine hydrochloride 96%
2-Nitrobenzylamine hydrochloride
Formel: C₇H₈N₂O₂·HCl
Molecular Weight: 188,62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00136280
CAS-Nummer: 24835-08-3