((Cyclopentyloxy)methyl)benzol 98%

Lieferant: Apollo Scientific

OR914680-1G OR914680-5G
APOSOR914680-1GEA 145 EUR
APOSOR914680-1G APOSOR914680-5G
((Cyclopentyloxy)methyl)benzol 98%
((Cyclopentyloxy)methyl)benzene
Formel: C₁₂H₁₆O
Lagertemperatur: Raumtemperatur
MDL: MFCD16295098
CAS-Nummer: 40843-99-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR