7-Methoxy-1H-indole-2,3-dione 95%
Lieferant: Apollo Scientific
Synonyme:
7-Methoxyindoline-2,3-dione
OR916226-5G
APOSOR916226-5GEA
71.2
EUR
APOSOR916226-5G
7-Methoxy-1H-indole-2,3-dione 95%
7-Methoxy-1H-indole-2,3-dione
Formel:
C₉H₇NO₃ Molecular Weight: 177,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00666831 CAS-Nummer: 84575-27-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...