[1,3-Bis(diphenylphosphino)propane]palladium(II) dichloride 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Dichloro[1,3-bis(diphenylphosphino)propane]palladium(II)

OR916291-5G OR916291-1G
APOSOR916291-5GEA 220 EUR
APOSOR916291-5G APOSOR916291-1G
[1,3-Bis(diphenylphosphino)propane]palladium(II) dichloride 98%
[1,3-Bis(diphenylphosphino)propane]palladium(II) dichloride
Formel: C₂₇H₂₆Cl₂P₂Pd
Molecular Weight: 589,77 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03844773
CAS-Nummer: 59831-02-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR