Benzyl-2-naphthyl ether 95%

Lieferant: Apollo Scientific

Synonyme: Benzyl 2-Naphthyl Ether, 2-Benzyloxynaphthalene

OR916445-100G OR916445-25G
APOSOR916445-100GEA 65.4 EUR
APOSOR916445-100G APOSOR916445-25G
Benzyl-2-naphthyl ether 95%
Benzyl-2-naphthyl ether
Formel: C₁₇H₁₄O
Molecular Weight: 234,30 g/mol
Schmelzpunkt: 96…98 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00089092
CAS-Nummer: 613-62-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR