(Ethoxymethyl)benzene 95%

Lieferant: Apollo Scientific

Synonyme: Benzyl ethyl ether

OR916482-5G
APOSOR916482-5GEA 92.8 EUR
APOSOR916482-5G
(Ethoxymethyl)benzene 95%
(Ethoxymethyl)benzene
Formel: C₉H₁₂O
Molecular Weight: 136,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00039903
CAS-Nummer: 539-30-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR