tert-Butylbenzoat 95%
Lieferant: Apollo Scientific
Synonyme:
Benzoic acid tert-butyl ester
OR916491-100G
APOSOR916491-100GEA
161
EUR
APOSOR916491-100G
tert-Butylbenzoat 95%
tert-Butylbenzoat
Formel:
C₁₁H₁₄O₂ Molecular Weight: 178,2277 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00048244 CAS-Nummer: 774-65-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...