Triethylsilanol 95%

Lieferant: Apollo Scientific

OR916626-25G OR916626-5G
APOSOR916626-25GEA 168 EUR
APOSOR916626-25G APOSOR916626-5G
Triethylsilanol 95%
Triethylsilanol
Formel: C₆H₁₆OSi
MDL: MFCD00042640
CAS-Nummer: 597-52-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR