Methyl 4,5-Dibromo-2-furancarboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: 4,5-Dibromo-2-furancarboxylic acid methyl ester, 4,5-Dibromo-2-furoic acid methyl ester, Methyl 4,5-Dibromo-2-furoate

OR916713-1G OR916713-5G
APOSOR916713-1GEA 66.7 EUR
APOSOR916713-1G APOSOR916713-5G
Methyl 4,5-Dibromo-2-furancarboxylate
Methyl 4,5-Dibromo-2-furancarboxylate
Formel: C₆H₄Br₂O₃
Molecular Weight: 283,90 g/mol
Schmelzpunkt: 56…57 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00092493
CAS-Nummer: 54113-41-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR