3-Chlorvanillin 95%

Lieferant: Apollo Scientific

OR916737-5G
APOSOR916737-5GEA 77 EUR
APOSOR916737-5G
3-Chlorvanillin 95%
3-Chlorvanillin
Formel: C₈H₇ClO₃
Molecular Weight: 186,59 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00016982
CAS-Nummer: 19463-48-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR