DL-Methioninmethylester Hydrochlorid 95%
Lieferant: Apollo Scientific
Synonyme: Methyl 2-amino-4-(methylthio)butanoate hydrochloride, H-DL-Met-OMe·HCl
OR916765-100G OR916765-25G OR916765-5G
APOSOR916765-100GEA 590 EUR
APOSOR916765-100G APOSOR916765-25G APOSOR916765-5G
DL-Methioninmethylester Hydrochlorid 95%
DL-Methioninmethylester Hydrochlorid
Formel: C₆H₁₃NO₂S·HCl
Molecular Weight: 199,7 g/mol
Schmelzpunkt: 110…117 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00067540
CAS-Nummer: 16118-36-8