2,2,4,4-Tetramethyl-3-pentanimin 95%
Lieferant: Apollo Scientific
Synonyme:
Di-tert-butylmethanimine, 2,2,4,4-Tetramethyl-3-pentanimine
OR916803-1G
OR916803-5G
APOSOR916803-1GEA
154
EUR
APOSOR916803-1G
APOSOR916803-5G
2,2,4,4-Tetramethyl-3-pentanimin 95%
2,2,4,4-Tetramethyl-3-pentanone imine
Formel:
C₉H₁₉N Molecular Weight: 141,26 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00216566 CAS-Nummer: 29097-52-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...