3-Methyl-1-phenyl-2-butanon 95%

Lieferant: Apollo Scientific

OR916904-25G OR916904-5G
APOSOR916904-25GEA 696 EUR
APOSOR916904-25G APOSOR916904-5G
3-Methyl-1-phenyl-2-butanon 95%
3-Methyl-1-phenyl-2-butanon
Formel: (C₆H₅)CH₂C(O)CH(CH₃)CH₃
Molecular Weight: 162,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00040487
CAS-Nummer: 2893-05-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR