(2-Methoxyphenyl)acetaldehyde 95%

Lieferant: Apollo Scientific

Synonyme: 2-(2-Methoxyphenyl)acetaldehyde

OR916906-1G OR916906-250MG
APOSOR916906-1GEA 217 EUR
APOSOR916906-1G APOSOR916906-250MG
(2-Methoxyphenyl)acetaldehyde 95%
(2-Methoxyphenyl)acetaldehyde
Formel: C₉H₁₀O₂
Molecular Weight: 150.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00072185
CAS-Nummer: 33567-59-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR