Ethyl α-chlorphenylacetat 95%
Lieferant: Apollo Scientific
Synonyme:
Ethyl 2-chloro-2-phenylacetate
OR916917-25G
APOSOR916917-25GEA
172
EUR
APOSOR916917-25G
Ethyl α-chlorphenylacetat 95%
Ethyl α-chlorphenylacetat
Formel:
C₁₀H₁₁ClO₂ Molecular Weight: 198,6461 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013675 CAS-Nummer: 4773-33-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...