6-Chloro-N-methyl-2-pyridinamine 98%
Lieferant: Apollo Scientific
Synonyme:
6-Chloro-N-methylpyridin-2-amine
OR917005-10G
OR917005-1G
OR917005-5G
APOSOR917005-10GEA
143
EUR
APOSOR917005-10G
APOSOR917005-1G
APOSOR917005-5G
6-Chloro-N-methyl-2-pyridinamine 98%
6-Chloro-N-methyl-2-pyridinamine
Formel:
C₆H₇ClN₂ Molecular Weight: 142,59 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234196 CAS-Nummer: 89026-78-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...