Tetrahydrofurfuryl butyrate 95%

Lieferant: Apollo Scientific

Synonyme: Butyric Acid Tetrahydrofurfuryl Ester, 2-(Butyryloxymethyl)tetrahydrofuran

OR917287-100G OR917287-25G
APOSOR917287-100GEA 502 EUR
APOSOR917287-100G APOSOR917287-25G
Tetrahydrofurfuryl butyrate 95%
Tetrahydrofurfuryl butyrate
Formel: C₉H₁₆O₃
Molecular Weight: 172,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00059734
CAS-Nummer: 2217-33-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR