(S)-(-)-1-Ethyl-2-pyrrolidinecarboxamide 95%
Lieferant: Apollo Scientific
Synonyme:
N-Ethyl-L-prolinamide
OR917343-1G
OR917343-250MG
APOSOR917343-1GEA
464
EUR
APOSOR917343-1G
APOSOR917343-250MG
(S)-(-)-1-Ethyl-2-pyrrolidinecarboxamide 95%
(S)-(-)-1-Ethyl-2-pyrrolidinecarboxamide
Formel:
C₇H₁₄N₂O Molecular Weight: 142,20 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191361 CAS-Nummer: 114812-34-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...