2-Chloro-5-methylbenzonitrile 95%
Lieferant: Apollo Scientific
OR917353-5G
APOSOR917353-5GEA
84.1
EUR
APOSOR917353-5G
2-Chloro-5-methylbenzonitrile 95%
2-Chloro-5-methylbenzonitrile
Formel:
C₈H₆ClN Lagertemperatur: Raumtemperatur |
MDL:
MFCD00045595 CAS-Nummer: 4387-32-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...