3-Chloro-5-methyl-1H-pyrazole 95%

Lieferant: Apollo Scientific

Synonyme: 5-Chloro-3-methyl-1H-pyrazole

OR917576-1G
APOSOR917576-1GEA 52.2 EUR
APOSOR917576-1G
3-Chloro-5-methyl-1H-pyrazole 95%
3-Chloro-5-methyl-1H-pyrazole
Formel: C₄H₅ClN₂
Molecular Weight: 116,55 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00233972
CAS-Nummer: 15953-45-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR