Acetoxime benzoate 95%

Lieferant: Apollo Scientific

Synonyme: Acetoxime Benzoate, O-Benzoyloxime 2-Propanone, Propan-2-one O-benzoyl oxime, O-Benzoylacetoxime

OR917748-25G
APOSOR917748-25GEA 177 EUR
APOSOR917748-25G
Acetoxime benzoate 95%
Acetoxime benzoate
Formel: C₁₀H₁₁NO₂
Molecular Weight: 177,20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00191697
CAS-Nummer: 942-89-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR