Propyl acetoacetate 95%

Lieferant: Apollo Scientific

Synonyme: Acetoacetic Acid Propyl Ester

OR917778-100G OR917778-25G
APOSOR917778-100GEA 371 EUR
APOSOR917778-100G APOSOR917778-25G
Propyl acetoacetate 95%
Propyl acetoacetate
Formel: C₇H₁₂O₃
Molecular Weight: 144,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00059405
CAS-Nummer: 1779-60-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR