(2E)-4-(4-Methylphenyl)-4-oxobut-2-enoic acid 95%

Lieferant: Apollo Scientific

Synonyme: 3-(4-Methylbenzoyl)acrylic acid

OR917829-1G OR917829-5G
APOSOR917829-1GEA 84.1 EUR
APOSOR917829-1G APOSOR917829-5G
(2E)-4-(4-Methylphenyl)-4-oxobut-2-enoic acid 95%
(2E)-4-(4-Methylphenyl)-4-oxobut-2-enoic acid
Formel: C₁₁H₁₀O₃
Molecular Weight: 190,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00075338
CAS-Nummer: 20972-36-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR