5-Methyl-2-(2,2,2-trimethylacetamido)pyridine 95%
Lieferant: Apollo Scientific
Synonyme:
N-(5-Methylpyridin-2-yl)pivalamide
OR917877-10G
OR917877-1G
OR917877-5G
APOSOR917877-10GEA
328
EUR
APOSOR917877-10G
APOSOR917877-1G
APOSOR917877-5G
5-Methyl-2-(2,2,2-trimethylacetamido)pyridine 95%
5-Methyl-2-(2,2,2-trimethylacetamido)pyridine
Formel:
C₁₁H₁₆N₂O Molecular Weight: 192,262 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03374646 CAS-Nummer: 86847-78-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...