((S)-Piperidin-2-yl)methanol 97%

Lieferant: Apollo Scientific

Synonyme: (S)-Piperidin-2-ylmethanol

OR917897-1G OR917897-5G
APOSOR917897-1GEA 128 EUR
APOSOR917897-1G APOSOR917897-5G
((S)-Piperidin-2-yl)methanol 97%
((S)-Piperidin-2-yl)methanol
Formel: C₆H₁₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD11036291
CAS-Nummer: 41373-39-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR