Methyl-3-methoxyacrylat 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 3-methoxyacrylate

OR917991-100G OR917991-500G
APOSOR917991-100GEA 75.4 EUR
APOSOR917991-100G APOSOR917991-500G
Methyl-3-methoxyacrylat 97%
Methyl-3-methoxyacrylat
Formel: C₅H₈O₃
Molecular Weight: 116,1152 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00128206
CAS-Nummer: 34846-90-7
UN: 3334
ADR: 9,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR