N-Methyl-1-(1-methylpiperidin-4-yl)methanamine 95%

Lieferant: Apollo Scientific

OR918101-100MG OR918101-1G OR918101-250MG
APOSOR918101-100MGEA 85.7 EUR
APOSOR918101-100MG APOSOR918101-1G APOSOR918101-250MG
N-Methyl-1-(1-methylpiperidin-4-yl)methanamine 95%
N-Methyl-1-(1-methylpiperidin-4-yl)methanamine
Formel: C₈H₁₈N₂
Molecular Weight: 142.25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06738983
CAS-Nummer: 405928-19-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR