(2-Chlorothiazol-5-yl)methylamine hydrochloride 98%
Lieferant: Apollo Scientific
Synonyme: (2-Chlorothiazol-5-yl)methanamine hydrochloride
OR918146-1G OR918146-25G OR918146-5G
APOSOR918146-1GEA 109 EUR
APOSOR918146-1G APOSOR918146-25G APOSOR918146-5G
(2-Chlorothiazol-5-yl)methylamine hydrochloride 98%
(2-Chlorothiazol-5-yl)methylamine hydrochloride
Formel: C₄H₆Cl₂N₂S
Molecular Weight: 185.08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09039253
CAS-Nummer: 153471-67-1