(S)-(+)-1-Benzyloxy-2-propanol 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: (S)-1-Benzyloxy-2-propanol, Benzyl (S)-(+)-2-Hydroxypropyl Ether

OR918206-25G OR918206-5G
APOSOR918206-25GEA 342 EUR
APOSOR918206-25G APOSOR918206-5G
(S)-(+)-1-Benzyloxy-2-propanol 95%
(S)-(+)-1-Benzyloxy-2-propanol
Formel: C₁₀H₁₄O₂
Molecular Weight: 166,22 g/mol
Dichte: 1,044 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD07367008
CAS-Nummer: 85483-97-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR