Methyl-2-amino-3-methoxybenzoate 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 2-amino-3-methoxybenzoate

OR918235-5G
APOSOR918235-5GEA 132 EUR
APOSOR918235-5G
Methyl-2-amino-3-methoxybenzoate 95%
Methyl-2-amino-3-methoxybenzoate
Formel: C₉H₁₁NO₃
Molecular Weight: 181,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11113109
CAS-Nummer: 5121-34-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR