2-(2,6-Dibromophenyl)acetic acid 98%

Lieferant: Apollo Scientific
Achtung

OR918350-1G OR918350-250MG
APOSOR918350-1GEA 66.7 EUR
APOSOR918350-1G APOSOR918350-250MG
2-(2,6-Dibromophenyl)acetic acid 98%
2-(2,6-Dibromophenyl)acetic acid
Formel: C₈H₆Br₂O₂
Molecular Weight: 293.94 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09832981
CAS-Nummer: 901310-02-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR