Methyl-3-thiophencarboxylat 98%
Lieferant: Apollo Scientific
Synonyme:
Thiophene-3-carboxylic Acid Methyl Ester
OR918386-100G
OR918386-25G
APOSOR918386-100GEA
258
EUR
APOSOR918386-100G
APOSOR918386-25G
Methyl-3-thiophencarboxylat 98%
Methyl-3-thiophencarboxylat
Formel:
C₆H₆O₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD06203810 CAS-Nummer: 22913-26-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...