7-Methylimidazo[1,2-a]pyridin-2-carbaldehyd 98%

Lieferant: Apollo Scientific

OR918559-1G OR918559-250MG
APOSOR918559-1GEA 451 EUR
APOSOR918559-1G APOSOR918559-250MG
7-Methylimidazo[1,2-a]pyridin-2-carbaldehyd 98%
7-Methylimidazo[1,2-a]pyridin-2-carbaldehyd
Formel: C₉H₈N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD09994888
CAS-Nummer: 202348-54-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR