9,9-Dimethylxanthene 98%
Lieferant: Apollo Scientific
Synonyme:
9,9-Dimethyl-9H-xanthene
OR918675-25G
OR918675-5G
OR918675-100G
APOSOR918675-25GEA
60.9
EUR
APOSOR918675-25G
APOSOR918675-5G
APOSOR918675-100G
9,9-Dimethylxanthene 98%
9,9-Dimethylxanthene
Formel:
C₁₅H₁₄O Molecular Weight: 210,28 g/mol Schmelzpunkt: 35…38 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00134434 CAS-Nummer: 19814-75-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...