2-Chlorcyclohexanon 95%

Lieferant: Apollo Scientific

OR918717-25G OR918717-5G
APOSOR918717-25GEA 91.5 EUR
APOSOR918717-25G APOSOR918717-5G
2-Chlorcyclohexanon 95%
2-Chlorcyclohexanon
Formel: C₆H₉ClO
Molecular Weight: 132,59 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00001626
CAS-Nummer: 822-87-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR