8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride 97%

Lieferant: Apollo Scientific

OR918784-1G OR918784-5G
APOSOR918784-1GEA 104 EUR
APOSOR918784-1G APOSOR918784-5G
8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride 97%
8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride
Formel: C₆H₁₂ClNO
Molecular Weight: 149,62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09800611
CAS-Nummer: 54745-74-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR