(Methylthio)acetaldehyddimethylacetal 95%
Lieferant: Apollo Scientific
Synonyme:
(Methylthio)acetaldehydedimethylacetal
OR918956-25G
APOSOR918956-25GEA
88.6
EUR
APOSOR918956-25G
(Methylthio)acetaldehyddimethylacetal 95%
(Methylthio)acetaldehyddimethylacetal
Formel:
C₅H₁₂O₂S Molecular Weight: 136,21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009848 CAS-Nummer: 40015-15-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...