N-Phenylurethan 95%

Lieferant: Apollo Scientific

OR919113-25G OR919113-100G
APOSOR919113-25GEA 132 EUR
APOSOR919113-25G APOSOR919113-100G
N-Phenylurethan 95%
N-Phenylurethan
Formel: C₉H₁₁NO₂
Molecular Weight: 165.19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00026806
CAS-Nummer: 101-99-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR