Quinoline-6-carboxamide 95%

Lieferant: Apollo Scientific

OR919127-1G
APOSOR919127-1GEA 209 EUR
APOSOR919127-1G
Quinoline-6-carboxamide 95%
Quinoline-6-carboxamide
Formel: C₁₀H₈N₂O
Molecular Weight: 172,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01312760
CAS-Nummer: 5382-43-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR