3-Benzotriazol-1-ylpropionic acid 96%

Lieferant: Apollo Scientific

OR919129-1G OR919129-5G
APOSOR919129-1GEA 161 EUR
APOSOR919129-1G APOSOR919129-5G
3-Benzotriazol-1-ylpropionic acid 96%
3-Benzotriazol-1-ylpropionic acid
Formel: C₉H₉N₃O₂
Molecular Weight: 191,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00964153
CAS-Nummer: 654-15-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR