4-Amino-5-(ethylsulphonyl)-2-methoxybenzoic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Amino-5-(ethylsulphonyl)-2-methoxybenzoic Acid
OR919210-1G
OR919210-25G
OR919210-5G
APOSOR919210-1GEA
14.7
EUR
APOSOR919210-1G
APOSOR919210-25G
APOSOR919210-5G
4-Amino-5-(ethylsulphonyl)-2-methoxybenzoic acid 98%
4-Amino-5-(ethylsulphonyl)-2-methoxybenzoic acid
Formel:
C₁₀H₁₃NO₅S Molecular Weight: 259,28 g/mol Schmelzpunkt: 82…85 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD04973619 CAS-Nummer: 71675-87-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...