5-Ethylisoxazole-3-carboxylic acid 96%
Lieferant: Apollo Scientific
OR919230-1G
OR919230-250MG
APOSOR919230-1GEA
256
EUR
APOSOR919230-1G
APOSOR919230-250MG
5-Ethylisoxazole-3-carboxylic acid 96%
5-Ethylisoxazole-3-carboxylic acid
Formel:
C₆H₇NO₃ Molecular Weight: 141.13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05864544 CAS-Nummer: 52320-59-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...