2-Chlorcycloheptanon 95%

Lieferant: Apollo Scientific

OR919389-1G OR919389-5G
APOSOR919389-1GEA 203 EUR
APOSOR919389-1G APOSOR919389-5G
2-Chlorcycloheptanon 95%
2-Chlorcycloheptanon
Formel: C₇H₁₁ClO
Lagertemperatur: Raumtemperatur
MDL: MFCD00041033
CAS-Nummer: 766-66-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR