2,6-Bis(chlormethyl)pyridin 95%
Lieferant: Apollo Scientific
OR919509-1G
OR919509-25G
OR919509-5G
APOSOR919509-1GEA
87
EUR
APOSOR919509-1G
APOSOR919509-25G
APOSOR919509-5G
2,6-Bis(chlormethyl)pyridin 95%
2,6-Bis(chloromethyl)pyridine
Formel:
C₇H₇Cl₂N Molecular Weight: 176,04 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00142844 CAS-Nummer: 3099-28-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...