8-Chinolinthiolhydrochlorid 95%
Lieferant: Apollo Scientific
Synonyme:
Thiooxine Hydrochloride, 8-Mercaptoquinoline Hydrochloride
OR919640-1G
OR919640-250MG
APOSOR919640-1GEA
91.5
EUR
APOSOR919640-1G
APOSOR919640-250MG
8-Chinolinthiolhydrochlorid 95%
8-Chinolinthiolhydrochlorid
Formel:
C₉H₇NS Molecular Weight: 197,68 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00043261 CAS-Nummer: 34006-16-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...