N-(4-Hydroxyphenyl)Methacrylamid 98%

Lieferant: Apollo Scientific

OR919641-25G OR919641-5G
APOSOR919641-25GEA 287 EUR
APOSOR919641-25G APOSOR919641-5G
N-(4-Hydroxyphenyl)Methacrylamid 98%
N-(4-Hydroxyphenyl)Methacrylamid
Formel: C₁₀H₁₁NO₂
Lagertemperatur: Raumtemperatur
CAS-Nummer: 19243-95-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR