2-(4-Methylphenoxy)ethyl-acetat 95%
Lieferant: Apollo Scientific
Synonyme:
2-(p-Tolyloxy)ethyl acetate, Ethylene Glycol Mono-p-tolyl Ether Acetate, Acetic Acid 2-p-Tolyloxyethyl Ester, 2-p-Tolyloxyethyl Acetate
OR919731-10G
OR919731-1G
OR919731-5G
APOSOR919731-10GEA
134
EUR
APOSOR919731-10G
APOSOR919731-1G
APOSOR919731-5G
2-(4-Methylphenoxy)ethyl-acetat 95%
2-(4-Methylphenoxy)ethyl-acetat
Formel:
C₁₁H₁₄O₃ Molecular Weight: 194,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00059337 CAS-Nummer: 6807-11-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...