1-(Methoxymethyl)benzotriazol 95%

Lieferant: Apollo Scientific

Synonyme: 1-(Methoxymethyl)-1H-benzotriazole

OR919800-1G OR919800-5G
APOSOR919800-1GEA 255 EUR
APOSOR919800-1G APOSOR919800-5G
1-(Methoxymethyl)benzotriazol 95%
1-(Methoxymethyl)benzotriazole
Formel: C₈H₉N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD00216630
CAS-Nummer: 71878-80-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR